Learn More
Waters Corp NW10 Centering Ring
Catalog No. 50792251 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
NW10 Centering Rings are essential components in vacuum technology, designed to provide a precise seal between two flanged connections. They typically ensure proper alignment of vacuum components and create a leak-tight connection when used with an O-ring.
  • Features common to NW10 Centering Rings include construction from materials such as aluminum or 304 stainless steel.
  • These rings are designed to accommodate various O-ring materials, including Buna-N (Nitrile NBR), Viton (Fluorocarbon, FKM), and Silicone.
  • O-ring materials typically support different maximum operating temperatures, for instance, Buna-N to 100°C, and Viton or Silicone up to 200°C.
Catalog No. 50-792-251 Supplier Waters Corp Supplier No. 6071200
NW10 Centering Rings are essential components in vacuum technology, designed to provide a precise seal between two flanged connections. They typically ensure proper alignment of vacuum components and create a leak-tight connection when used with an O-ring.
  • Features common to NW10 Centering Rings include construction from materials such as aluminum or 304 stainless steel.
  • These rings are designed to accommodate various O-ring materials, including Buna-N (Nitrile NBR), Viton (Fluorocarbon, FKM), and Silicone.
  • O-ring materials typically support different maximum operating temperatures, for instance, Buna-N to 100°C, and Viton or Silicone up to 200°C.