Learn More
TARGETMOL CHEMICALS INC NX-2127 1MG
SDP
Catalog No. 502877931
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. NX-2127 (ETX2514 Triethylamine) is an orally active BTK inhibitor that induces degradation of mutant BTKC481S in cells.NX-2127 has potent antiproliferative activity and inhibits the proliferation of BTKC481S mutant TMD8 cells.NX-2127 potently catalyzes the degradation of Ikaros (IKZF1) and Aiolos (IKZF3) at 25 nM and 54 nM, respectively. NX-2127 is associated with the immune system, stimulating T cell activation and increasing IL-2 production in primary human T cells. Purity 98.11%
Catalog No. 50-287-7931 Supplier TARGETMOL CHEMICALS INC Supplier No. T734631MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. NX-2127 (ETX2514 Triethylamine) is an orally active BTK inhibitor that induces degradation of mutant BTKC481S in cells.NX-2127 has potent antiproliferative activity and inhibits the proliferation of BTKC481S mutant TMD8 cells.NX-2127 potently catalyzes the degradation of Ikaros (IKZF1) and Aiolos (IKZF3) at 25 nM and 54 nM, respectively. NX-2127 is associated with the immune system, stimulating T cell activation and increasing IL-2 production in primary human T cells. Purity 98.11%