Learn More
Selleck Chemical LLC NX-2127-E1381-5MG
SDP
Catalog No. 503136976
Encompass_Preferred
NX-2127 is a unique potent inhibitor of BTK that prevents its functions by catalyzing the ubiquitylation and proteasomal degradation of BTK rather than via direct binding NX-2127 stimulates T cell activation and increases IL-2 production in primary human T Cells
Catalog No. 50-313-6976 Supplier Selleck Chemical LLC Supplier No. E13815MG
NX-2127 is a unique potent inhibitor of BTK that prevents its functions by catalyzing the ubiquitylation and proteasomal degradation of BTK rather than via direct binding NX-2127 stimulates T cell activation and increases IL-2 production in primary human T Cells