Learn More
MARSHALL SCIENTIFIC Edwards nXDS20i Dry Scroll Pump
SDP
Catalog No. NC2888778
Encompass_Preferred
With exceptional pumping capability, ultimate vacuum performance and state-of-the-art design features Edwards nXDS is the best performing pump in its class. nXDS improves legacy XDS pumps by offering increased pumping speeds, combined with lower ultimate pressures, lower power consumption and lower noise - up to 20 times quieter than other pumps. Gas ballast allows for pumping of condensable vapours including water, solvents, dilute acids, and bases. nXDS pumps also feature the latest in tip seal technology giving significantly longer life between tip seal changes. Integrated inverter drive with auto sensing voltage input delivers optimized pumping performance globally. Ultimate vacuum of 0.030 mbar and peak pumping speed of 13.0 CFM.
Catalog No. NC2888778 Supplier MARSHALL SCIENTIFIC Supplier No. EDA73801983
Add to Cart
Add to Cart
Request a Quote
With exceptional pumping capability, ultimate vacuum performance and state-of-the-art design features Edwards nXDS is the best performing pump in its class. nXDS improves legacy XDS pumps by offering increased pumping speeds, combined with lower ultimate pressures, lower power consumption and lower noise - up to 20 times quieter than other pumps. Gas ballast allows for pumping of condensable vapours including water, solvents, dilute acids, and bases. nXDS pumps also feature the latest in tip seal technology giving significantly longer life between tip seal changes. Integrated inverter drive with auto sensing voltage input delivers optimized pumping performance globally. Ultimate vacuum of 0.030 mbar and peak pumping speed of 13.0 CFM.