Learn More
Aviva Systems Biology Corp NXF1 Antibody - N-terminal region (ARP40639_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40639_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NXF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human NXF1. Alternate names: TAP, MEX67ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57563