Learn More
Aviva Systems Biology Corp NXF5 Antibody - middle region (ARP41062_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41062_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NXF5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NXF5. Alternate names: ApplicationIHC, WB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57706