Learn More
LGC GENOMICS LLC (Lucigen) NxGen phi29 DNA Polymerase, 2,000 U @ 10 U/LExcellent strand displacement, fidelity and speed.
Catalog No. NC2160348
Encompass
Supplied at 10,000 U/mL in 10 mM Tris-HCl, 100 mM KCl, 0.1 mM EDTA, 1 mM dithiothreitol, 0.5% Tween-20, 0.5% NP-40, 50% glycerol, pH 7.4 @ 25C. Also supplied is 10X phi29 DNA Polymerase Buffer composed of 500 mM Tris-HCl, 100 mM (NH4)2SO4, 40 mM Dithiothreitol, 100 mM MgCl2, pH 7.5 @ 25C. 10,000 U is supplied as 5 2,000 U.
This item is not returnable. View return policy
Catalog No. NC2160348 Supplier LGC GENOMICS LLC Supplier No. 302211
Supplied at 10,000 U/mL in 10 mM Tris-HCl, 100 mM KCl, 0.1 mM EDTA, 1 mM dithiothreitol, 0.5% Tween-20, 0.5% NP-40, 50% glycerol, pH 7.4 @ 25C. Also supplied is 10X phi29 DNA Polymerase Buffer composed of 500 mM Tris-HCl, 100 mM (NH4)2SO4, 40 mM Dithiothreitol, 100 mM MgCl2, pH 7.5 @ 25C. 10,000 U is supplied as 5 2,000 U.