Learn More
LGC GENOMICS LLC (Lucigen) NxGen phi29 DNA Polymerase, 2,000 U @ 10 U/LExcellent strand displacement, fidelity and speed.
Catalog No. NC1793942
Encompass
phi29 DNA Polymerase is a highly processive DNA polymerase with a powerful strand displacement activity and a 3 5 proofreading exonuclease function. The enzyme is capable of up to 70,000 insertions per binding event.
Catalog No. NC1793942 Supplier LGC GENOMICS LLC Supplier No. 302212
phi29 DNA Polymerase is a highly processive DNA polymerase with a powerful strand displacement activity and a 3 5 proofreading exonuclease function. The enzyme is capable of up to 70,000 insertions per binding event.