Learn More
LGC GENOMICS LLC (Lucigen) NxGen RNAse Inhibitor, 10,000 U @ 40 U/LEliminate RNase activity.
Catalog No. NC1819281
Encompass
RNAse Inhibitor is a potent inhibitor of ribonucleases such as RNase A, RNase B, and RNase C. The 52 kDa protein is a fusion of the porcine RNAse Inhibitor gene with a proprietary 22.5 kDa protein tag.Supplied at 40,000 U/mL in 20 mM HEPES-KOH, 50 mM KCl, 8 mM dithiothreitol, 50% glycerol, pH 7.5 @ 25¿C.
Catalog No. NC1819281 Supplier LGC GENOMICS LLC Supplier No. 302811
RNAse Inhibitor is a potent inhibitor of ribonucleases such as RNase A, RNase B, and RNase C. The 52 kDa protein is a fusion of the porcine RNAse Inhibitor gene with a proprietary 22.5 kDa protein tag.Supplied at 40,000 U/mL in 20 mM HEPES-KOH, 50 mM KCl, 8 mM dithiothreitol, 50% glycerol, pH 7.5 @ 25¿C.