Learn More
LGC GENOMICS LLC (Lucigen) NxGen T4 DNA Ligase (Low Concentration)A workhorse, high quality DNA ligase for joining blunt or cohesive ends of dsDNA or dsRNA.
Supplier:  LGC GENOMICS LLC 302411
Encompass
NxGen T4 DNA Ligase (Low Concentration)A workhorse, high quality DNA ligase for joining blunt or cohesive ends of dsDNA or dsRNA.Reliably join double-stranded nucleic acid chains.The low concentration (LC) NxGen T4 DNA Ligase Kit includes T4 DNA Ligase at 2 U/¿L and 10X T4 DNA ligase Buffer. The high concentration(HC) NxGen T4 DNA Ligase Kit includes NxGen T4 DNA Ligase at 10 U/¿L and both 2X Rapid Ligation Buffer and 10X T4 DNA ligase Buffer.
Catalog No. NC1967390