Learn More
Cayman Chemical Annexin V APC Assay Kit, 1 ea
SDP
Catalog No. 502896971
Encompass_Preferred
Cayman's Annexin V APC Assay Kit employs an APC-conjugated annexin V as a probe for phosphatidylserine on the outer membrane of apoptotic cells. DAPI is used as a marker of cell membrane permeability seen in very late apoptotic or necrotic cells. This kit is valuable for researchers looking to assess the degree of apoptosis induced by experimental compounds, and can be useful in pharmacology, immunology, cancer, or physiology research.
Catalog No. 50-289-6971 Supplier Cayman Chemical Supplier No. 6014101 EA
Cayman's Annexin V APC Assay Kit employs an APC-conjugated annexin V as a probe for phosphatidylserine on the outer membrane of apoptotic cells. DAPI is used as a marker of cell membrane permeability seen in very late apoptotic or necrotic cells. This kit is valuable for researchers looking to assess the degree of apoptosis induced by experimental compounds, and can be useful in pharmacology, immunology, cancer, or physiology research.