Learn More
Aviva Systems Biology Corp NXN Antibody - N-terminal region (ARP61611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NXN. It was validated on Western Blot. . Alternate names: FLJ12614, NRX, TRG-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-420