Learn More
Aviva Systems Biology Corp NXNL1 Antibody - N-terminal region (ARP65360_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65360_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NXNL1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human NXNL1. Alternate names: RDCVF, TXNL6ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-288-485