Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10093MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: His. Host: Mammal. Species Reactivity: Human. Protein Expression: A DNA sequence encoding the Homo sapiens (Human) Neurexophilin-2, was expressed in the hosts and tags indicated. AA Sequence: KEVVHATEGL DWEDKDAPGT LVGNVVHSRI ISPLRLFVKQ SPVPKPGPMA YADSMENFWD WLANITEIQE PLARTKRRPI VKTGKFKKMF GWGDFHSNIK TVKLNLLITG KIVDHGNGTF SVYFRHNSTG LGNVSVSLVP PSKVVEFEVS PQSTLETKES KSFNCRIEYE KTDRAKKTAL CNFDPSKICY QEQTQSHVSW LCSKPFKVIC IYIAFYSVDY KLVQKVCPDY NYHSETPYLS SG. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: NXPH2. Gene ID: 11249. Uniprot: O95156.
Catalog No. 50-188-5493