Learn More
Aviva Systems Biology Corp NXPH3 Antibody - N-terminal region (ARP52305_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52305_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NXPH3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NXPH3. Alternate names: NPH3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-696