Learn More
LGC GENOMICS LLC NxSeq AmpFREE Low DNA Library Kit 48 reactions
Catalog No. NC3355911
Encompass
The NxSeq AmpFREE Low DNA Library Kit is compatible with our NxSeq Adaptors or other dT-tailed, Illumina-compatible adaptors.Each NxSeq AmpFREE Low DNA Library Kit contains Enzyme Mix (EM), 2X Buffer (2XB), Ligase (LIG) and Elution Buffer (EB). Adaptors must be purchased separately.
Catalog No. NC3355911 Supplier LGC GENOMICS LLC Supplier No. 140002
The NxSeq AmpFREE Low DNA Library Kit is compatible with our NxSeq Adaptors or other dT-tailed, Illumina-compatible adaptors.Each NxSeq AmpFREE Low DNA Library Kit contains Enzyme Mix (EM), 2X Buffer (2XB), Ligase (LIG) and Elution Buffer (EB). Adaptors must be purchased separately.