Learn More
Aviva Systems Biology Corp NXT1 Antibody - N-terminal region (ARP54919_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54919_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NXT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NXT1. Alternate names: MTR2, P15ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-976