Learn More
Medchemexpress LLC HY-114263 10mg Medchemexpress, NXT629 CAS:1454925-59-7 Purity:>98%
SDP
Catalog No. 501969315
Encompass_Preferred
Medchemexpress, HY-114263 10mg NXT629 CAS:1454925-59-7 NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9315 Supplier Medchemexpress LLC Supplier No. HY11426310MG
Medchemexpress, HY-114263 10mg NXT629 CAS:1454925-59-7 NXT629 is a potent, selective, and competitive PPAR-α antagonist, with an IC50 of 77 nM for human PPARα, shows high selectivity over other nuclear hormone receptor, such as PPARδ, PPARγ, ERβ, GR and TRβ, IC50s are 6.0, 15, 15.2, 32.5 and >100 μM, respectively. NXT629 has potent anti-tumor activity and inhibits experimental metastasis of cancer cell in animal models. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.