Learn More
Cayman Chemical Nexturastat A, Purity: ≥95%, CAS: 1403783-31-2, 10 mg
SDP
Catalog No. 502965008
Encompass_Preferred
A potent, selective inhibitor of HDAC6 (IC50 = 5.02 nM); dose-dependently induces hyperacetylation of α-tubulin in B16 murine melanoma cells without elevating histone H3 acetylation
Catalog No. 50-296-5008 Supplier Cayman Chemical Supplier No. 1687410 MG
A potent, selective inhibitor of HDAC6 (IC50 = 5.02 nM); dose-dependently induces hyperacetylation of α-tubulin in B16 murine melanoma cells without elevating histone H3 acetylation