Learn More
Selleck Chemical LLC NXY-059 (Disufenton sodium) 10mg 168021-79-2 Cerovive, Disufenton Sodium
SDP
Supplier:  Selleck Chemical LLC S600210MG
Encompass_Preferred
NXY-059 (Cerovive, Disufenton Sodium) is a novel nitrone, shows efficacious neuroprotective effects. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-686-3