Learn More
Cole-Parmer Cole-Parmer Nylon Syringe Filters, 0.45 um, 25 mm Dia; 100/Box
Supplier:  Cole-Parmer 0291514
Encompass
Filters are Triton-free and have a bidirectional membrane support and luer lock inlet/luer slip outlet for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. All Cole-Parmer syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. Sterile filters are sterilized by gamma radiation. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
Catalog No. NC0061709