Learn More
Sterlitech Corporation Nylon Syringe Filters, Sterlitech, 0.22 Micron, 25mm, 100/Pk
SDP
Catalog No. 501952320
Encompass_Preferred
Nylon Syringe Filter Applications:* HPLC aqueous and organic sample filtration* Solvent filtration and clarification* Exceptionally low extractable level with no wetting agents utilized* Filter viscous aqueous and organic HPLC sample filtration* Improve sample volume throughput with prefilter* High particulate load
Catalog No. 50-195-2320 Supplier Sterlitech Corporation Supplier No. 1470349
Add to Cart
Add to Cart
Nylon Syringe Filter Applications:* HPLC aqueous and organic sample filtration* Solvent filtration and clarification* Exceptionally low extractable level with no wetting agents utilized* Filter viscous aqueous and organic HPLC sample filtration* Improve sample volume throughput with prefilter* High particulate load