Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accurate Chemical Nycodenz Density Gradient Medium, Powder, Off-white, 500 g
SDP

Catalog No. NC1878833
Encompass

Nycodenz 500GM is a universal, non-ionic, and non-toxic density gradient medium designed for the high-resolution separation of biological particles. It comes as an off-white powder that forms stable low-osmolarity solutions. This product is ideal for isolating various cellular components without affecting their viability. Nycodenz has minimal interference with biochemical assays, making it an excellent choice for researchers seeking reliable separation methods.

  • Form: Powder
  • Color: Off-white
  • Chemical Name: Iohexol
  • Molecular Weight: 821 Da
  • Density (Solid): 2.1 g/ml
  • Maximum Solution Density: 1.426 g/ml (at 80% w/v)
  • Solubility: Water, formamide, dimethyl formamide
  • Stability: Up to 5 years
  • Packaging Size: 500 g

Catalog No. NC1878833 Supplier Accurate Chemical Supplier No. AN1002424
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nycodenz 500GM is a universal, non-ionic, and non-toxic density gradient medium designed for the high-resolution separation of biological particles. It comes as an off-white powder that forms stable low-osmolarity solutions. This product is ideal for isolating various cellular components without affecting their viability. Nycodenz has minimal interference with biochemical assays, making it an excellent choice for researchers seeking reliable separation methods.

  • Form: Powder
  • Color: Off-white
  • Chemical Name: Iohexol
  • Molecular Weight: 821 Da
  • Density (Solid): 2.1 g/ml
  • Maximum Solution Density: 1.426 g/ml (at 80% w/v)
  • Solubility: Water, formamide, dimethyl formamide
  • Stability: Up to 5 years
  • Packaging Size: 500 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.