Learn More
Crescent Chemical Co Inc NYCODENZ 60% (W/V)
SDP
Catalog No. 50247952
Encompass_Preferred
Nycodenz , 60 % (w/v) solution in water Non-ionic density gradient medium, similar to the former Metrizamide but less toxic. Mammalian cells and viruses isolated in Nycodenz gradients potentially retain better functional integrity than in Metrizamide gradients. Nycodenz is suitable f
Catalog No. 50-247-952 Supplier Crescent Chemical Co Inc Supplier No. SE31000.01
Nycodenz , 60 % (w/v) solution in water Non-ionic density gradient medium, similar to the former Metrizamide but less toxic. Mammalian cells and viruses isolated in Nycodenz gradients potentially retain better functional integrity than in Metrizamide gradients. Nycodenz is suitable f