Learn More
CELLTREAT Scientific Products Nylon Syringe Filter, 0.22m, 30mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229761
Encompass_Preferred
CELLTREAT Scientific Products 0.22¿m Nylon Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. Nylon. Universal application for analytical procedures. Chemical filtration. Beverage filtration. Membrane diameter 30mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister pack. 30/case.
Catalog No. 50-403-870