Learn More
CELLTREAT Scientific Products Nylon Syringe Filter, 0.45m, 13mm, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229754
Encompass_Preferred
CELLTREAT Scientific Products 0.45¿m Nylon Syringe Filter. Filters are molded in the polypropylene housings, providing a reliable seal without the use of adhesives. Female luer-lock inlet and make luer slip outlet. Color coded by membrane type for easy identification. Nylon. Universal application for analytical procedures. Chemical filtration. Beverage filtration. Membrane diameter 13mm. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped in peel-to-open paper/plastic blister packs. 75/case.
Catalog No. 50-202-069