Learn More
MSC Tilsatec Cut and Abrasion-Resistant Gloves, GP2000MF, Polyester, Microfoam Nitrile, Small, 12/Pk.
Catalog No. 5000688266
Encompass
These cut and abrasion-resistant gloves provide essential hand protection for industrial tasks. Constructed from a durable polyester knit, they feature a micro-foam nitrile coating on the palms and fingertips to ensure superior grip in various conditions. Designed with a seamless lining, these unisex gloves offer a comfortable fit and consistent performance across a range of applications.
  • ANSI/ISEA cut resistance level A1
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 0
  • Micro-foam nitrile coated palms and fingertips
  • Knit cuff style
  • Seamless polyester lining
  • Red glove color with black coating
Catalog No. 50-006-88266 Supplier MSC Supplier No. 89738199
These cut and abrasion-resistant gloves provide essential hand protection for industrial tasks. Constructed from a durable polyester knit, they feature a micro-foam nitrile coating on the palms and fingertips to ensure superior grip in various conditions. Designed with a seamless lining, these unisex gloves offer a comfortable fit and consistent performance across a range of applications.
  • ANSI/ISEA cut resistance level A1
  • ANSI/ISEA abrasion resistance level 3
  • ANSI/ISEA puncture resistance level 0
  • Micro-foam nitrile coated palms and fingertips
  • Knit cuff style
  • Seamless polyester lining
  • Red glove color with black coating