Learn More
Component Supply NYL MESH FILT,70 MICR,49 X1YD
SDP
Supplier:  Component Supply UCMN70
Encompass_Preferred
Nylon filter mesh, plain weave, 70 micron mesh opening, 49 percent open area, 1 lineal yard
Catalog No. NC0601590