Learn More
MSC Carlisle Nylon Paddle Scraper, 48 in, Green
Supplier:  MSC 95753653
Encompass
This durable paddle scraper is made from sturdy nylon, designed for everyday mixing and stirring tasks in various applications. Its unique blade contours and shapely corners ensure access to hard-to-reach areas, while its sealed exterior allows for easy cleaning. The long handle is ideal for deep pots and vats, and a hanging tab provides sanitary storage.
  • Durable sturdy nylon for everyday mixing and stirring tasks.
  • Contouring blade sharpens and contours to specific applications with prolonged use.
  • Shapely corners provide access to hard-to-reach bowl corners.
  • Color-coded to reduce cross-contamination and promote food safety.
  • Long handle extends into deep pots or vats.
  • Sealed exterior is smooth and easy to clean.
  • Hanging tab facilitates sanitary storage on hooks or cord.
Catalog No. 50-284-0022