Learn More
Electron Microscopy Sciences Nylon Tip Paint Markers, 1/EA
SDP
Catalog No. NC1878881
Encompass_Preferred
The marker quickly dispenses fast drying enamel from a fine point. These pens have nylon tips and work on all types of surfaces including metal, glass, plastic, wood and paper. They are resilient to heat and lead free. Color: Black Color: Black.
Catalog No. NC1878881 Supplier Electron Microscopy Sciences Supplier No. 6205366
The marker quickly dispenses fast drying enamel from a fine point. These pens have nylon tips and work on all types of surfaces including metal, glass, plastic, wood and paper. They are resilient to heat and lead free. Color: Black Color: Black.