Learn More
MSC PIP Utility Gloves, Pigskin Leather Palm, Small, 1/Pr.
Catalog No. 5000684755
Encompass
These size small brown gloves feature a top grain pigskin leather palm and a breathable fabric back, designed for durability and comfort. The construction includes a slip-on cuff and hook and loop closure for a secure fit.
  • Top grain pigskin leather palm construction provides breathability, softens with use, and withstands moisture without stiffening
  • Reinforced thumb crotch offers extended glove life, enhanced comfort, and increased abrasion resistance in a high-wear area
  • Pre-curved fingers improve comfort and reduce hand fatigue
  • Four-way stretch thumb provides increased flexibility and comfort
  • Hook and loop closure allows for easy on/off and a secure fit
  • Ultra-breathable back keeps hands cool and comfortable
  • Performance rated EN 388 3122X with ANSI puncture level 3
Catalog No. 50-006-84755 Supplier MSC Supplier No. 14160378
These size small brown gloves feature a top grain pigskin leather palm and a breathable fabric back, designed for durability and comfort. The construction includes a slip-on cuff and hook and loop closure for a secure fit.
  • Top grain pigskin leather palm construction provides breathability, softens with use, and withstands moisture without stiffening
  • Reinforced thumb crotch offers extended glove life, enhanced comfort, and increased abrasion resistance in a high-wear area
  • Pre-curved fingers improve comfort and reduce hand fatigue
  • Four-way stretch thumb provides increased flexibility and comfort
  • Hook and loop closure allows for easy on/off and a secure fit
  • Ultra-breathable back keeps hands cool and comfortable
  • Performance rated EN 388 3122X with ANSI puncture level 3