Learn More
Selleck Chemical LLC Nylidrin Hydrochloride S5267-25mg
SDP
Catalog No. 503112726
Encompass_Preferred
Nylidrin Hydrochloride (Buphenine) a -adrenergic agonist is used therapeutically for its vasodilating effect on the peripheral circulation and possibly on the cerebral circulation as well
Catalog No. 50-311-2726 Supplier Selleck Chemical LLC Supplier No. S526725MG
Nylidrin Hydrochloride (Buphenine) a -adrenergic agonist is used therapeutically for its vasodilating effect on the peripheral circulation and possibly on the cerebral circulation as well