Learn More
Medchemexpress LLC Nylidrin hydrochloride | 849-55-8 | 98.7% | 335.87 g·mol⁻¹ | C19H26ClNO2 | 10 MG
SDP
Catalog No. 5000298882
Encompass_Preferred
Nylidrin hydrochloride is the hydrochloride salt of buphenine, a research-grade β-adrenergic agonist with reported NMDA NR1A/2B antagonism and antiviral activity against H1N1 influenza. It is used in receptor pharmacology and preclinical models of hemorrhagic shock and allergic responses.
  • Orally active β-adrenergic agonist for receptor pharmacology studies.
  • Reported NMDA NR1A/2B antagonist activity.
  • Demonstrated antiviral activity against multiple H1N1 influenza A subtypes.
  • Used in hemorrhagic shock and anti-allergic research models.
  • High-purity research reagent suitable for analytical and in vivo studies.
Catalog No. 50-002-98882 Supplier Medchemexpress LLC Supplier No. HYB140410MG
Nylidrin hydrochloride is the hydrochloride salt of buphenine, a research-grade β-adrenergic agonist with reported NMDA NR1A/2B antagonism and antiviral activity against H1N1 influenza. It is used in receptor pharmacology and preclinical models of hemorrhagic shock and allergic responses.
  • Orally active β-adrenergic agonist for receptor pharmacology studies.
  • Reported NMDA NR1A/2B antagonist activity.
  • Demonstrated antiviral activity against multiple H1N1 influenza A subtypes.
  • Used in hemorrhagic shock and anti-allergic research models.
  • High-purity research reagent suitable for analytical and in vivo studies.