Learn More
Medchemexpress LLC Nylidrin hydrochloride | 849-55-8 | 98.7% | 5 MG
SDP
Catalog No. 5000411882
Encompass_Preferred
Nylidrin hydrochloride, also known as Buphenine hydrochloride, is an orally active β-adrenergic agonist. This compound demonstrates significant biological activity, including antagonism of NR1A/2B NMDA receptors, reduction of NP, HA, and M1 levels, and antiviral efficacy against H1N1 influenza A viruses. It also shows promise in improving hemorrhagic shock and exhibiting anti-allergic effects.
  • Orally active β-adrenergic agonist
  • Antagonizes NR1A/2B NMDA receptors (IC50 = 0.18 μM in Xenopus oocytes)
  • Reduces levels of NP, HA, and M1
  • Demonstrates antiviral activity against multiple H1N1 subtype influenza A viruses
  • Improves hemorrhagic shock
  • Possesses anti-allergic effects
Catalog No. 50-004-11882 Supplier Medchemexpress LLC Supplier No. HYB14045MG
Nylidrin hydrochloride, also known as Buphenine hydrochloride, is an orally active β-adrenergic agonist. This compound demonstrates significant biological activity, including antagonism of NR1A/2B NMDA receptors, reduction of NP, HA, and M1 levels, and antiviral efficacy against H1N1 influenza A viruses. It also shows promise in improving hemorrhagic shock and exhibiting anti-allergic effects.
  • Orally active β-adrenergic agonist
  • Antagonizes NR1A/2B NMDA receptors (IC50 = 0.18 μM in Xenopus oocytes)
  • Reduces levels of NP, HA, and M1
  • Demonstrates antiviral activity against multiple H1N1 subtype influenza A viruses
  • Improves hemorrhagic shock
  • Possesses anti-allergic effects