Learn More
IW Tremont Co Inc LabExact Hydrophilic Nylon membrane, NON-STERILE, 0.22 micron, 47mm diameter, pack of 100
SDP
Supplier:  IW Tremont Co Inc LEIWTNY022047
Encompass_Preferred
LabExact Nylon Membrane is supported with a polyethylene terephthalate (PET) scrim for added support. This scrim is bonded to the nylon using a unique technique with no bonding agent for purity, chemical compatibility and lower extractables. Hydrophilic material eliminates need for wetting agents. Strong protein bonding with high recoveries
Catalog No. 50-223-7598