Learn More
IW Tremont Co Inc LabExact Hydrophilic Nylon membrane, NON-STERILE, 0.45 micron, 47mm diameter, pack of 100
SDP
Catalog No. 502237602
Encompass_Preferred
LabExact Nylon Membrane is supported with a polyethylene terephthalate (PET) scrim for added support. This scrim is bonded to the nylon using a unique technique with no bonding agent for purity, chemical compatibility and lower extractables. Hydrophilic material eliminates need for wetting agents. Strong protein bonding with high recoveries
This item is not returnable. View return policy
Catalog No. 50-223-7602 Supplier IW Tremont Co Inc Supplier No. LEIWTNY045047
LabExact Nylon Membrane is supported with a polyethylene terephthalate (PET) scrim for added support. This scrim is bonded to the nylon using a unique technique with no bonding agent for purity, chemical compatibility and lower extractables. Hydrophilic material eliminates need for wetting agents. Strong protein bonding with high recoveries