Learn More
Waters Corp Waters Corporation Syringe Filter, Nylon, 13 mm, 0.2 μm
Supplier:  Waters Corp WAT200524
Encompass_Preferred
This hydrophilic membrane works well with both aqueous and organic solvents. Nylon is particularly suitable for high-pH samples but should be avoided in any protein recovery applications.
  • Hydrophilic membrane suitable for both aqueous and organic solvents.
  • Nylon suitable for high-pH samples.
  • Ensures superior filtration for analytical samples.
  • HPLC-certified for low extractables.
  • Polypropylene support with hydrophilic nylon membrane.
  • Prevents “air bubble” lock with 13 mm small spike outlet.
  • Provides low extractable materials for less chromatographic interference.
Catalog No. 50-786-070