Learn More
Waters Corp Waters Acrodisc Syringe Filter, Nylon, 13 mm, 0.2 μm, Aqueous
Supplier:  Waters Corp WAT200834
Encompass_Preferred
This filter features a hydrophilic nylon membrane with a polypropylene support, designed for high-quality filtration of analytical samples. It is ideal for life science, pharmaceutical methods, research and development, quality control, or specialty environmental applications. Certified for HPLC, it ensures low extractables for reliable results.
  • 13 mm diameter, 0.2 μm pore size
  • 1000 units per case
  • Handles aqueous and organic solvents; suitable for high-pH samples
  • < 30 μL hold-up volume, < 10 mL sample volume
  • Low extractables and API adsorption for analytical analysis
  • Prevents “air bubble” lock with a 13 mm mini spike outlet
Catalog No. 50-786-165