Learn More
Waters Corp Waters Acrodisc Syringe Filter, Nylon, 13 mm, 0.45 μm, Aqueous
Supplier:  Waters Corp WAT200832
Encompass_Preferred
The Nylon Acrodisc Syringe Filter is designed to streamline sample preparation with its versatile hydrophilic membrane. This filter is suitable for a wide range of applications, especially those involving high-pH samples. It comes in a convenient 1000-case package, ensuring ample supply for laboratory needs.
  • Hydrophilic membrane, compatible with both aqueous and organic solvents.
  • Ideal for use with high-pH samples.
  • Not recommended for protein recovery applications.
Catalog No. 50-786-163