Learn More
Waters Corp Waters Syringe Filter, Nylon, 25 mm, 0.45 μm
Supplier:  Waters Corp WAT200518
Encompass_Preferred
This hydrophilic membrane works well with both aqueous and organic solvents. It is particularly suitable for high-pH samples but should be avoided in any protein recovery applications.
  • Works well with both aqueous and organic solvents.
  • Suitable for high-pH samples.
Catalog No. 50-786-064