Learn More
Waters Corp Waters Acrodisc Minispike Syringe Filter, Nylon, 13 mm, 0.2 μm
Supplier:  Waters Corp WAT200562
Encompass_Preferred
The Nylon Acrodisc Minispike Syringe Filter simplifies extraction and filtration of aqueous and organic solvents. HPLC-certified for low extractables, its hydrophilic nylon membrane in polypropylene housing is suitable for aqueous/organic liquids. A 13 mm spike outlet prevents “air bubble” lock when filling vials.
  • Simplifies extraction and filtration of aqueous and organic solvents.
  • HPLC-certified for low extractables.
  • Hydrophilic nylon membrane in polypropylene housing.
  • 13 mm spike outlet prevents “air bubble” lock in vials.
  • Precise pore size ratings enhance chromatography and instrument protection.
  • Features lower extractables, reduced API adsorption, and less chromatographic interference.
  • Homogenous membrane materials ensure consistent analytical findings.
Catalog No. 50-786-097