Learn More
Mopec Nylon Arson Evidence Bags
SDP
Catalog No. 22446183 Shop All Mopec Products
Change view
Click to view available options
Dimensions (L x W):
12 x 18 in.
18 x 36 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-446-183 12 x 18 in.
22-446-070 18 x 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-183 Supplier Mopec Supplier No. FS054
Add to Cart
Add to Cart
Designed for secure storage of materials containing accelerants, allowing easy viewing and labeling.
  • Can be sealed with cable ties in the field or a heat sealer in the lab
  • Made of Nylon
  • Allows marking with standard pens
  • Prevents loss of flammables and hydrocarbons
  • Suitable for Long-term, stackable storage
Specifications
Dimensions (L x W) 12 x 18 in.
Product Type Evidence Bag
Color Transparent
Quantity 20/Pk.