Learn More
Electron Microscopy Sciences Nylon Biopsy Bag, 98 x 75 mm
SDP
Catalog No. 5030339
Encompass_Preferred
Biopsy bags made from nylon cloth, 0.2 mm mesh diameter. All seams are heat-sealed. Allows for unrestricted fluid movement around the tissue. Reduces the loss of small biopsy specimens. Reduces cross contamination and pressure artifacts by using sponges.
Size: 98 x 75 mm
Catalog No. 50-303-39 Supplier Electron Microscopy Sciences Supplier No. 6232498
Biopsy bags made from nylon cloth, 0.2 mm mesh diameter. All seams are heat-sealed. Allows for unrestricted fluid movement around the tissue. Reduces the loss of small biopsy specimens. Reduces cross contamination and pressure artifacts by using sponges.
Size: 98 x 75 mm