Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Nylon Biopsy Bags

Catalog No. 6774009 Shop All Epredia Products
Change view
Click to view available options
Size:
Small
Medium
Large
Quantity:
1000/Pk.
200/Pk.
500/Pk.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity
6774009 Medium 1000/Pk.
6774010 Small 1000/Pk.
6774011 Large 500/Pk.
6774012 Medium 200/Pk.
6774013 Medium 500/Pk.
6774014 Small 200/Pk.
6774015 Small 500/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 6774009 Supplier Epredia™ Supplier No. 6774009
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure unrestricted fluid movement around tissue with Epredia™ Nylon Biopsy Bags.

  • Reduce risk of small specimen loss
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)

Specifications

Product Type Biopsy Bag
Material Nylon
Color Blue
Length (Metric) 45 mm
Width (Metric) 65 mm
Length (English) 1.8 in.
Width (English) 2.6 in.
Size Medium
Quantity 1000/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.