Learn More
Med Vet International Nylon Brush, Wire Handle, 12 in.
Catalog No. 502829753
Encompass_Preferred
Made for cleaning funnel stems and capillary tubes. White nylon bristles 1/2? x 2 1/2? long. Overall length: 12?
Catalog No. 50-282-9753 Supplier Med Vet International Supplier No. LB09
Made for cleaning funnel stems and capillary tubes. White nylon bristles 1/2? x 2 1/2? long. Overall length: 12?