Learn More
GUARDIAN EQUIPMENT NYLON DECK FLANGE
Catalog No. NC1318230
Encompass_Preferred
Nylon deck flange assembly, consisting of: 350-011G; Nylon deck flange with 1' NPS thread, 350-011-1; O-ring, 840-08LN-R; 1" NPS aluminum locknut and washer (pkg of 2)
Catalog No. NC1318230 Supplier GUARDIAN EQUIPMENT Supplier No. AP350011
Nylon deck flange assembly, consisting of: 350-011G; Nylon deck flange with 1' NPS thread, 350-011-1; O-ring, 840-08LN-R; 1" NPS aluminum locknut and washer (pkg of 2)