Learn More
Med Vet International Jor-Vet Nylon Muzzle, J0169R, Set of 7
Catalog No. 5000495333
Encompass_Preferred
Nylon Muzzle. JorVet introduces an expanded line of high quality water resistant nylon muzzles that have a side easy-release snap buckle. Comes in a variety of sizes. Features:Safety. Muzzles fit securely with adjusting straps for a snug fit.Comfort. Lightweight nylon feels and looks more professional than gauze roll.Quality. Reinforced stitching to assure long-lasting use.
Catalog No. 50-004-95333 Supplier Med Vet International Supplier No. JOR169R
Nylon Muzzle. JorVet introduces an expanded line of high quality water resistant nylon muzzles that have a side easy-release snap buckle. Comes in a variety of sizes. Features:Safety. Muzzles fit securely with adjusting straps for a snug fit.Comfort. Lightweight nylon feels and looks more professional than gauze roll.Quality. Reinforced stitching to assure long-lasting use.