Learn More
MSC Electro Hardware Cap Screw, Nylon, 3/8-16 x 3/4 in, 100/Pack
Catalog No. 502867703
Encompass
This Hex Head Cap Screw features a 3/8-16 thread size with a 3/4 in length under the head. It is made from Grade 6/6 Nylon and has a bright/uncoated finish, suitable for various applications requiring corrosion resistance and electrical insulation.
  • Hex head design
  • Threaded 3/8-16 UNC
  • Length under head is 3/4 in
  • Constructed from durable Grade 6/6 Nylon
  • Bright/uncoated finish
  • Fully threaded
  • Compatible with 9/16 in hex key
  • Meets ASTM D789-66 Type 1 and GDE2 standards
Catalog No. 50-286-7703 Supplier MSC Supplier No. 70998935
This Hex Head Cap Screw features a 3/8-16 thread size with a 3/4 in length under the head. It is made from Grade 6/6 Nylon and has a bright/uncoated finish, suitable for various applications requiring corrosion resistance and electrical insulation.
  • Hex head design
  • Threaded 3/8-16 UNC
  • Length under head is 3/4 in
  • Constructed from durable Grade 6/6 Nylon
  • Bright/uncoated finish
  • Fully threaded
  • Compatible with 9/16 in hex key
  • Meets ASTM D789-66 Type 1 and GDE2 standards