Learn More
Tisch Scientific Tisch Scientific Nylon Membrane Filter, 0.45 µm, 47 mm, Sterile, 200 Pack, SF18013
SDP
Catalog No. NC2079913
Encompass
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.
Catalog No. NC2079913 Supplier Tisch Scientific Supplier No. SF18013
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.