Learn More
Tisch Scientific Tisch Scientific Nylon Membrane Filter, 0.45 µm, 47 mm, Nonsterile, 200 Pack, SF13880
SDP
Catalog No. NC2076739
Encompass
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.
Catalog No. NC2076739 Supplier Tisch Scientific Supplier No. SF13880
Tisch nylon membrane filters are naturally hydrophilic, have high non-specific binding properties, and an extractable level of <0.0015 mg/cm^2, they are ideal for HPLC solvent and sample preparation.